Methyl arachidate 99%
Glass bottle, 1 g
Supplier: Thermo Fisher Scientific
Methyl arachidate 99%
Catalog # ACRO452820010
Supplier: Thermo Fisher Scientific
CAS Number: 1120-28-1
Jump to:
Product Details & Documents
Synonyms: Methyl eicosanoate, Methyl Arachidate
- CAS Number:1120-28-1
- Formula:C₂₁H₄₂O₂
- MDL Number:MFCD00009014
- Molecular Weight:326.56 g/mol
- Boiling Pt:215…216 °C (10 mmHg)
- Melting Pt:45…48 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:45 to 48 °C
- GC:≥98.5 %
- Free acid:≤1 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



