Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Agar, Bacteriological Grade

Agar, Bacteriological Grade

Plastic bottle, 250 g
Catalog # ACRO443572500
Agar, Bacteriological Grade
Catalog # ACRO443572500
CAS Number:  9002-18-0
Agar, Bacteriological Grade
Agar, Bacteriological Grade

Plastic bottle, 250 g

CAS Number:  9002-18-0
Price
SFr. 93.50
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Melting Pt:90 °C
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    White to cream to light yellow to beige Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤15 % (1 g, 105°C)
  • Solubility:
    (1.5 % in hot water) Clear to slightly opalescent light yellow to yellow solution
  • Gel strength:
    ≥700 g/cm² (1.5 %)
  • Gel point:
    32 to 38°C (1.5 %)
  • Ash:
    ≤5 %
  • pH:
    6.0 to 7.5 (10 % suspension)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.