Cyclohexanone 99+% ACS
Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Molecular Weight:98.14 g/mol
- Boiling Pt:155 °C (760 mmHg)
- Melting Pt:–47 °C
- Density:0.947 g/cm³
- Flash Pt:43 °C
- Storage Temperature:Ambient
- UN:1915
- ADR:3,III
Documents
Specifications
Specification Test Results
- Appearance:Clear viscous liquid
- Water:≤0.05 %
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.05 %
Specifications
- Pack type:Glass bottle
- Pk:250 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



