Cyclohexanone 99+% ACS
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Cyclohexanone 99+% ACS
Catalog # 406090025.
Supplier: Thermo Fisher Scientific
CAS Number: 108-94-1
Jump to:
Product Details & Documents
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Molecular Weight:98.14 g/mol
- Boiling Pt:155 °C (760 mmHg)
- Melting Pt:–47 °C
- Density:0.947 g/cm³
- Flash Pt:43 °C
- Storage Temperature:Ambient
- UN:1915
- ADR:3,III
Documents
Specifications
Specification Test Results
- Appearance:Clear viscous liquid
- Water:≤0.05 %
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.05 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Cat. No.:406090025.
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



