Order Entry
Switzerland
Orders LinkContactUsLinkComponent
(2-Hydroxylpropyl)-β-cyclodextrine 97%

(2-Hydroxylpropyl)-β-cyclodextrine 97%

Glass bottle, 500 g
Catalog # ACRO297565000
(2-Hydroxylpropyl)-β-cyclodextrine 97%
Catalog # ACRO297565000
CAS Number:  128446-35-5
(2-Hydroxylpropyl)-β-cyclodextrine 97%
(2-Hydroxylpropyl)-β-cyclodextrine 97%

Glass bottle, 500 g

CAS Number:  128446-35-5
Price
SFr. 1’470.00
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
  • CAS Number:128446-35-5
  • Formula:C₆₃H₁₁₂O₄₂
  • MDL Number:MFCD16621721
  • Melting Pt:278 °C (dec.)
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤7.5 % (K.F.)
  • HPLC:
    ≥96.0 % (On dry substance)
  • NMR:
    2 to 6 (degree of substitution)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.