Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Hydrocortisone 98%

Hydrocortisone 98%

Glass bottle, 100 g
Catalog # ACRO352451000
Health
Hydrocortisone 98%
Catalog # ACRO352451000
CAS Number:  50-23-7
Hydrocortisone 98%
Hydrocortisone 98%

Glass bottle, 100 g

CAS Number:  50-23-7
Price
1 870.00 SFr.
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:50-23-7
  • Formula:C₂₁H₃₀O₅
  • MDL Number:MFCD00011654
  • Molecular Weight:362.47 g/mol
  • Boiling Pt:567 °C (1013 hPa)
  • Melting Pt:212…217 °C
  • Density:1,4 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-020-1

Documents

Specifications

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    212 to 217 °C
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +150° to +156° (20 °C, 589 nm) (C=1, dioxane)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.