Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Ethidium bromide 95%, pure

Glass bottle, 25 g
Catalog # ACRO170960250
Toxic
Health
Ethidium bromide 95%, pure
Catalog # ACRO170960250
CAS Number:  1239-45-8
Ethidium bromide 95%, pure
Ethidium bromide 95%, pure

Glass bottle, 25 g

CAS Number:  1239-45-8
Price
SFr. 781.00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Merck Index:13,04751
  • Boiling Pt:72 °C (0,1 -rr)
  • Melting Pt:260…262 °C (dec.)
  • Density:1,01 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:214-984-6
  • UN:2811
  • ADR:6.1,I

 

Specifications

Specification Test Results

  • Appearance:
    Red to dark purple Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥94 %
  • Loss on drying:
    ≤5.0 %
  • Solubility:
    (1 % in water) Clear red
  • pH:
    (1 % in water at 20°C) Approx. 4.5

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications