5α-Cholestane 98+%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:481-21-0
- Formula:C₂₇H₄₈
- MDL Number:MFCD00066412
- Molecular Weight:372.68 g/mol
- Merck Index:12,02254
- Boiling Pt:250 °C (1 mmHg)
- Melting Pt:78…81 °C
- Density:0,9133 g/cm³ (80 °C)
- :Density: 0.9133 g/cm³ (80 °C)
- Storage Temperature:Ambient
- EINECS:207-562-8
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:78 to 81 °C
- GC:≥98.0 %
- Specific optical rotation:+23° to +30° (20 °C, 589 nm) (c=2, CHCl3)
Specifications
- Pack type:Glass bottle
- Pk:5 g



