Silver sulphate ACS
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Silver sulphate ACS
Catalog # ACRO194071000
Supplier: Thermo Fisher Scientific
CAS Number: 10294-26-5
Jump to:
Product Details & Documents
Synonyms: Disilver sulphate
- CAS Number:10294-26-5
- Formula:Ag₂SO₄
- MDL Number:MFCD00003407
- Molecular Weight:311.8 g/mol
- Boiling Pt:1085 °C (760 mmHg)
- Melting Pt:652 °C
- Density:5,45 g/cm³ (20 °C)
- Storage Temperature:Ambient
- UN:3077
- ADR:9,III
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Titration Argentometric:≥98.0 %
- Nitrate (NO3):≤0.001 %
- Iron (Fe):≤0.001 %
- Insoluble matter:≤0.02 % (in water and AgCl)
- Subst not pptd by HCl:≤0.03 %
Specifications
- Pack type:Glass bottle
- Pk:100 g



