Vanadium(V) oxide 98+%
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Vanadium(V) oxide 98+%
Catalog # ACRO207990025
Supplier: Thermo Fisher Scientific
CAS Number: 1314-62-1
Jump to:
Product Details & Documents
Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
- CAS Number:1314-62-1
- Formula:O₅V₂
- MDL Number:MFCD00011457
- Molecular Weight:181.88 g/mol
- Merck Index:13,09987
- Boiling Pt:1750 °C (760 mmHg)
- Melting Pt:690 °C
- Density:3.35 g/cm³
- Storage Temperature:Ambient
- EINECS:215-239-8
- UN:2862
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:Yellow to orange-brown Powder
- Assay:≥98.0 %
- Particle size:≤500 µm
- Chromium (Cr):≤200 ppm
- Iron (Fe):≤1000 ppm
- Silicon (Si):≤200 ppm
- Sodium oxide (Na2O):≤300 ppm
- Potassium oxide (K2O):≤300 ppm
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg



