Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D-(+)-Mannose 99+%

D-(+)-Mannose 99+%

Plastic drum, 2,5 kg
Catalog # ACRO150600025
D-(+)-Mannose 99+%
Catalog # ACRO150600025
CAS Number:  3458-28-4
D-(+)-Mannose 99+%
D-(+)-Mannose 99+%

Plastic drum, 2,5 kg

CAS Number:  3458-28-4
Price
2 130.00 SFr.
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,05772
  • Boiling Pt:376 °C (1013 hPa)
  • Melting Pt:133 °C
  • Density:1,539 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:222-392-4

Documents

Specifications

Specification Test Results

  • Appearance:
    White Powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.45 %
  • HPLC:
    ≥99.0 %
  • Appearance of solution:
    (10 % in water) Clear
  • Bacterial endotoxins:
    (LAL-test) 0.5 E.U. in 1 ml of a 0.1% soln.
  • Specific optical rotation:
    +13.5° to +14.9° (20°C, 589 nm) (c=10, water) (after 24 hours, on dry substance)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    2,5 kg

Specifications