Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Pyrocatechol 99+%

Pyrocatechol 99+%

Plastic bottle, 2,5 kg
Catalog # ACRO158980025
Corrosive
Toxic
Health
Pyrocatechol 99+%
Catalog # ACRO158980025
CAS Number:  120-80-9
Pyrocatechol 99+%
Pyrocatechol 99+%

Plastic bottle, 2,5 kg

CAS Number:  120-80-9
Price
478.00 SFr.
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
  • CAS Number:120-80-9
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002188
  • Molecular Weight:110.11 g/mol
  • Merck Index:13,08089
  • Boiling Pt:245 °C (760 mmHg)
  • Melting Pt:103…106 °C
  • Density:1,0548 g/cm³ (20 °C)
  • Flash Pt:131 °C
  • Storage Temperature:Ambient
  • EINECS:204-427-5
  • UN:2811
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Off-white to grey-brown Crystalline flakes (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Melting point:
    103 to 106 °C
  • GC:
    ≥99.0 %
  • Heavy metals (as Pb):
    ≤10 ppm
  • Insoluble matter:
    ≤0.05 % (in water)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg

Specifications