Caprinaldehyde 95%
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Decylaldehyde, Decanal, Decyl aldehyde
- CAS Number:112-31-2
- Formula:C₁₀H₂₀O
- MDL Number:MFCD00007031
- Molecular Weight:156.27 g/mol
- Boiling Pt:207…209 °C (760 mmHg)
- Melting Pt:7 °C
- Density:0.83 g/cm³
- Flash Pt:85 °C
- Storage Temperature:Ambient
- EINECS:203-957-4
Documents
ACRO15497
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4270 to 1.4320 (20°C, 589 nm)
- GC:≥94 %
- NMR (1H-NMR):Consistent with structure
Specifications
- Pack type:Glass bottle
- Pk:500 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



