Cyclohexanone 99.8%, extra pure
Metal drum, 25 L
Supplier: Thermo Fisher Scientific
Cyclohexanone 99.8%, extra pure
Catalog # ACRO111190251
Supplier: Thermo Fisher Scientific
CAS Number: 108-94-1
Jump to:
Product Details & Documents
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Molecular Weight:98.14 g/mol
- Merck Index:13,02755
- Boiling Pt:155 °C (760 mmHg)
- Melting Pt:–47 °C
- Density:0.947 g/cm³
- Flash Pt:43 °C
- Storage Temperature:Ambient
- EINECS:203-631-1
- UN:1915
- ADR:3,III
- REACH:01-2119453616-35
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4450 to 1.4520 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥99.75 %
- Color scale:≤20 APHA
- Iron (Fe):≤0.001 %
- Magnesium (Mg):≤0.005 %
- Manganese (Mn):≤0.002 %
- Potassium (K):≤0.01 %
- Sodium (Na):≤0.05 %
- Insoluble matter:≤0.005 %
- Nitrogen compounds (as N):≤0.002 %
Specifications
- Pack type:Metal drum
- Pk:25 L
- Cat. No.:ACRO111190251
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



