4-Nitro-o-anisidine 99%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
4-Nitro-o-anisidine 99%
Catalog # ACRO150645000
Supplier: Thermo Fisher Scientific
CAS Number: 97-52-9
Jump to:
Product Details & Documents
Synonyms: 2-Methoxy-4-nitroaniline, 2-Amino-5-nitroanisole
- CAS Number:97-52-9
- Formula:C₇H₈N₂O₃
- MDL Number:MFCD00007363
- Molecular Weight:168.15 g/mol
- Boiling Pt:364 °C (1013 hPa)
- Melting Pt:139…142 °C
- Density:1,2112 g/cm³ (156 °C)
- Storage Temperature:Ambient
- EINECS:202-588-6
Documents
ACRO15064
SDS
Specifications
Specification Test Results
- Appearance:Yellow to orange Crystalline powder
- Infrared spectrum:Conforms
- Melting point:139 to 142 °C
- GC:≥98.5 %
Specifications
- Pack type:Plastic bottle
- Pk:500 g



