BD® Blunt Needles
18 G, 40 mm, Blunt bevel, Sterile, Red
Supplier: BD Medical
Jump to:
Product Details & Documents
BD blunt fill and blunt filter needles are specifically designed to reduce the risk of needlestick injuries during medication preparation.
- The blunt fill needle is designed for vial access and available with a BD Luer-Lok™ connection
- The blunt filter is designed for safe ampoule medication preparation
- BD blunt fill needles have female luer fittings, which mate to male luer fittings
- BD blunt filter needle has a 5-micron filter for particles such as glass and plastic fragments, for safer medication delivery
- Natural rubber latex is not part of the material formulation
- Sterile, singe use
: Note: The BD blunt fill and filter needles are not for skin injection.
Specifications
- Colour code:Red
- Gauge:18 G
- Type:Sterile
- Type of tip:Blunt bevel
- Length:40 mm
- Seq:03