Chromatography data station, DataApex ClarityVA
Colibrick A/D Converter 2 Channel
906-0195
ClarityVA is a flexible and powerful Chromatography Data System (CDS) that controls HPLC and GC systems. This modular software includes a lot of controls for many different systems, modules and interfaces. The flexibility enables an easy control of HPLC systems in combination with a user friendly surface.
- Direct control of Hitachi HPLC (Chromaster and Primaide)
- Modules for specific calculation and methodology
- Tools for regulated environment
- Extended user support
- Language localisations (English, German, French,...)
ClarityVA is useable for modern complained pharma labs, when configured correctly (21 CFR part 11 compliance).
- Description:Colibrick A/D Converter 2 Channel
- Seq:17