Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Precision butyrometer, without stopper

Precision butyrometer, without stopper

Butyrometer, precision, 0 - 4 % : 0,05, with conformity certificate
Catalog # GERB3150-C
Supplier:   GERBER FUNKE
Precision butyrometer, without stopper
Catalog # GERB3150-C
Supplier:  GERBER FUNKE
Precision butyrometer, without stopper

Butyrometer, precision, 0 - 4 % : 0,05, with conformity certificate

Price
27.70 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

For drinking vat milk, frosted rear scale wall, error tolerance 0,025%.

Specifications
  • Description:
    Butyrometer, precision, 0 - 4 % : 0,05, with conformity certificate
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Stopper, rubber, conical, for all measuring method butyrometers
    Stopper, rubber, conical, for all measuring method butyrometers
    614-2028
    Log in for availability
    1 * 100 ST - SFr. 44.90