Filter discs, DURAN®
1, 55 mm, 4,2 mm
Catalog # SCOT251555108
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.
- Available in intermediate sizes, alternative dimensions, alternate porosities and bi-planar design
Made from clear DURAN® borosilicate glass 3,3.
Specifications
- Ø:55 mm
- Porosity:1
- Seq:27
- Thickness:4,2 mm



