ContactUsLinkComponent
LESAFFRE LYNSIDE NUTRI OPTIMUM BA10 IS A BAKER'S INACTIVATED DRIEDYEAST, OBTAINED BY DRUM DRYING OF SACCHAROMYCES CEREVISIAE, PRIMARYGROWN ON A MOLASSES BASED MEDIA. 1 * 1 kg
Catalog # XXXX10738
Supplier:  Chemicals
LESAFFRE LYNSIDE NUTRI OPTIMUM BA10 IS A BAKER'S INACTIVATED DRIEDYEAST, OBTAINED BY DRUM DRYING OF SACCHAROMYCES CEREVISIAE, PRIMARYGROWN ON A MOLASSES BASED MEDIA. 1 * 1 kg
Catalog # XXXX10738
Supplier:  Chemicals
Frequently Bought Together