Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Filter discs, DURAN®

4, 55 mm, 4,2 mm
  SCOT251555408
Filter discs, DURAN®
  SCOT251555408
Filter discs, DURAN®
Filter discs, DURAN®

4, 55 mm, 4,2 mm

 
SFr. 23.20
   
1 * 1 ST
 

Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.

  • Available in intermediate sizes, alternative dimensions, alternate porosities and bi-planar design

Made from clear DURAN® borosilicate glass 3,3.

 
  • Ø:
    55 mm
  • Porosity:
    4
  • Seq:
    78
  • Thickness:
    4,2 mm