Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bottles, square

Bottles, square

100 ml, Bottles, square, RD 40, Clear, 46×46×71 mm
Catalog # RIXI9-2200-0100-40
Bottles, square
Catalog # RIXI9-2200-0100-40
Bottles, square

100 ml, Bottles, square, RD 40, Clear, 46×46×71 mm

Price
922.00 SFr.
Log in for availability
1 * 480 ST
Product Details & Documents

Square bottles with clear-sighted thread.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • Thread:
    RD 40
  • Capacity:
    100 ml
  • Colour:
    Clear
  • Description:
    Bottles, square
  • Seq:
    01
  • W×D×H:
    46×46×71 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.