ContactUsLinkComponent
Large Knitted Polyester Swab Cleanroom laundered double layer 100% polyester knit swab 20 Swabs / pack , 25pks / case price / case 1 * 500 items
  BERKLTP125S.25
 :  BERKSHIRE
Large Knitted Polyester Swab Cleanroom laundered double layer 100% polyester knit swab 20 Swabs / pack , 25pks / case price / case 1 * 500 items
  BERKLTP125S.25
 :  BERKSHIRE
Frequently Bought Together