Orders LinkContactUsLinkComponent
Phase separating filters
Phase separating filters
108 H, 240 mm
Catalog # PRATA240108H
Supplier:   PRAT DUMAS
Phase separating filters
Catalog # PRATA240108H
Supplier:  PRAT DUMAS
Phase separating filters
Phase separating filters
108 H, 240 mm
Price
SFr. 196.00
Log in for availability
1 * 100 ST
Product Details & Documents
Hydrophobic paper for the fast separation of aqueous solutions from hydrofuge solvents.
Useful as well with organic solvents heavier or lighter than water replacing many times bulbs to be settled.
Delivery information: Filters supplied as flat in boxes with 100 filters or folded in 4 by 100.
Specifications
  • Grade:
    108 H
  • Seq:
    06
  • Size:
    240 mm