Orders LinkContactUsLinkComponent
Phase separating filters
Phase separating filters
108 H, 90 mm
  PRATA090108H
 :   PRAT DUMAS
Phase separating filters
  PRATA090108H
Phase separating filters
Phase separating filters
108 H, 90 mm
SFr. 53.80
1 * 100 ST
Hydrophobic paper for the fast separation of aqueous solutions from hydrofuge solvents.
Useful as well with organic solvents heavier or lighter than water replacing many times bulbs to be settled.
Delivery information: Filters supplied as flat in boxes with 100 filters or folded in 4 by 100.
  • Grade:
    108 H
  • Seq:
    01
  • Size:
    90 mm