Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Digital block heaters

Digital block heaters

Block heater, digital, 1, 200 W, RT* +5...130 °C, ≤±1 °C, 288×200×100 mm, 2,9 kg
Catalog # 460-0306
Digital block heaters
Catalog # 460-0306
Digital block heaters
Digital block heaters

Block heater, digital, 1, 200 W, RT* +5...130 °C, ≤±1 °C, 288×200×100 mm, 2,9 kg

Price
623.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Versatile digital block heaters offer a wide range of blocks to accommodate various tubes provide temperature uniformity and stability for reproducible results. The units have digital controls and display of time and temperature and are durable with a powder-coated steel body. Ideal for sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation. Choice of models to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.

  • Built-in temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables user to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Interchangeable aluminum alloy sample blocks provide versatility and allow for easy cleaning and disinfecting

Delivery information: Supplied with block extraction tool, but without blocks, these must be ordered separately.

Caution: * RT = Ambient
Note: Temp. stability value shown above is at 37 °C.

Specifications
  • Heating capacity:
    200 W
  • No. of blocks:
    1
  • Temperature Range:
    RT* +5...130 °C
  • Weight:
    2,9 kg
  • Seq:
    01
  • W×D×H:
    288×200×100 mm
  • Temp. stability:
    ≤±1 °C
  • Type:
    Block heater, digital
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • PT1000 temperature probe
    PT1000 temperature probe
    460-0312
    Log in for availability
    1 * 1 ST - SFr. 47.80

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0301
    50 ml tubes
    4
    Log in for availability
    1 * 1 ST - SFr. 257.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0299
    0,5 ml + 0,2 ml Ø tubes
    30+20
    Log in for availability
    1 * 1 ST - SFr. 215.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0300
    15 ml tubes
    15
    Log in for availability
    1 * 1 ST - SFr. 257.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0298
    25 mm + 12-13 mm + 6 mm Ø tubes
    3+12+6
    Log in for availability
    1 * 1 ST - SFr. 215.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0297
    1,5 ml + 2,0 ml tubes
    18+10
    Log in for availability
    1 * 1 ST - SFr. 157.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Mixed aluminium block
    Mixed aluminium block
    460-0296
    6 mm + 10 mm Ø tubes
    32+21
    Log in for availability
    1 * 1 ST - SFr. 157.00

Product Information

  • Catalog #
    For
    W×D×H
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0295
    96-well microplate
    124×76×39 mm
    Log in for availability
    1 * 1 ST - SFr. 111.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0294
    0,5 ml tubes
    40
    Log in for availability
    1 * 1 ST - SFr. 131.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0293
    25 mm Ø tubes
    6
    Log in for availability
    1 * 1 ST - SFr. 121.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0292
    20 mm Ø tubes
    8
    Log in for availability
    1 * 1 ST - SFr. 121.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0291
    18 mm Ø tubes
    12
    Log in for availability
    1 * 1 ST - SFr. 149.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0290
    16 mm Ø tubes
    15
    Log in for availability
    1 * 1 ST - SFr. 149.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0289
    13 mm Ø tubes
    24
    Log in for availability
    1 * 1 ST - SFr. 125.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0288
    2,0 ml tubes
    28
    Log in for availability
    1 * 1 ST - SFr. 111.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0287
    1,5 ml tubes
    28
    Log in for availability
    1 * 1 ST - SFr. 111.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0286
    10 mm Ø tubes
    28
    Log in for availability
    1 * 1 ST - SFr. 121.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0285
    6 mm Ø tubes
    46
    Log in for availability
    1 * 1 ST - SFr. 121.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0305
    1×96 well PCR plate, half or full skirted
    -
    Log in for availability
    1 * 1 ST - SFr. 332.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0304
    1×96-well PCR plate, non skirted
    -
    Log in for availability
    1 * 1 ST - SFr. 289.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0303
    50 ml conical bottom tubes
    4
    Log in for availability
    1 * 1 ST - SFr. 521.00

Product Information

  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Aluminium block
    Aluminium block
    460-0302
    15 ml conical bottom tubes
    15
    Log in for availability
    1 * 1 ST - SFr. 521.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.