Modular storage systems, KP
KP-BLOCK, modular and stackable block storage metal cabinet with 13 drawers, red - capacity: 700 blocks
Catalog # 720-2810
Supplier: KLINIPATH
Jump to:
Product Details & Documents
Stackable, universal storage system for preparations and blocks. A complete system with growth potential. A modular system able to transform local storage in a laboratory into a long-term storage system. A growth system with easy exchangeable and removable plastic trays.
- Lower gliding resistance, no rusting because of paint damage
Specifications
- Description:KP-BLOCK, modular and stackable block storage metal cabinet with 13 drawers, red - capacity: 700 blocks
Accessories
Product Information
- Catalog #AvailabilityPrice
- Top for modular systems, KP Glass and KP BlockTop for modular systems, KP Glass and KP Block720-2813Log in for availability1 * 1 ST - SFr. 9.00
Product Information
- Catalog #AvailabilityPrice
- Base for modular systems, KP Glass and KP BlockBase for modular systems, KP Glass and KP Block720-2812Log in for availability1 * 1 ST - SFr. 59.70