ContactUsLinkComponent
Blade, trapezoid no.61329, deep-edged 2-sided ground-edge, non-corrosive blade, 2-times usable ,length:60mm, width:19mm, material th ickness:0.9mm, double bevel edge-ground. 1 * 10 items
Catalog # MARO61329.50
Supplier:  MARTOR ARGENTAX
Blade, trapezoid no.61329, deep-edged 2-sided ground-edge, non-corrosive blade, 2-times usable ,length:60mm, width:19mm, material th ickness:0.9mm, double bevel edge-ground. 1 * 10 items
Catalog # MARO61329.50
Supplier:  MARTOR ARGENTAX
Frequently Bought Together