Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Broad range hydrometers

Broad range hydrometers

1,000 − 1,500 °Bé, 0,005, 235 mm
Catalog # ALLA3050FG100/20QP
Supplier:   Alla France
Broad range hydrometers
Catalog # ALLA3050FG100/20QP
Supplier:  Alla France
Broad range hydrometers
Broad range hydrometers

1,000 − 1,500 °Bé, 0,005, 235 mm

Price
SFr. 41.10
Log in for availability
1 * 1 ST
Product Details & Documents

These broad range hydrometers are calibrated at 20 °C.

Mostly dedicated to applications with citric and tartaric acids.

Specifications
  • Length:
    235 mm
  • Seq:
    004
  • Division:
    0,005
  • Range:
    1,000 − 1,500 °Bé