Order Entry
Switzerland
Orders LinkContactUsLinkComponent
NORMAL HUMAN SERUM COMPLEMENT 1 * 5 ml

NORMAL HUMAN SERUM COMPLEMENT 1 * 5 ml

Catalog # BMDCA113.5ML
Marketsource
Supplier:   Biomedica
NORMAL HUMAN SERUM COMPLEMENT 1 * 5 ml
Catalog # BMDCA113.5ML
Supplier:  Biomedica
Marketsource
NORMAL HUMAN SERUM COMPLEMENT 1 * 5 ml
Price
SFr. 408.00
Log in for availability
1 * 5 mL
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.