ContactUsLinkComponent
PEQGREEN 20 µL SAMPLE DNA/RNA Dye Safe alternative for ethidium bromide for staining of dsDNA, ssDNA and RNA. - High level sensitivity, comparable to ethidium bromide - 20 000x concentrated in water - Direct addition to gel solution or staining after 1 * 20 µl
Catalog # PEQL37-5099
Supplier:  VWR Peqlab
PEQGREEN 20 µL SAMPLE DNA/RNA Dye Safe alternative for ethidium bromide for staining of dsDNA, ssDNA and RNA. - High level sensitivity, comparable to ethidium bromide - 20 000x concentrated in water - Direct addition to gel solution or staining after 1 * 20 µl
Catalog # PEQL37-5099
Supplier:  VWR Peqlab