Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cryoboxes

Cryoboxes

without grid insert, -, Tissue culture tubes 15/50 ml, 122×122×128 mm
Catalog # NEOL2-7094
Supplier:   NEOLAB
Cryoboxes
Catalog # NEOL2-7094
Supplier:  NEOLAB
Cryoboxes
Cryoboxes

without grid insert, -, Tissue culture tubes 15/50 ml, 122×122×128 mm

Price
SFr. 14.20
Log in for availability
1 * 1 ST
Product Details & Documents

Cryo resistant cardboard.

  • Water repellent
  • Llid has a marking area on one of the sides

For storage of tissue culture tubes of 15 and 50 ml capacities for cryogenic deep freezers.

Specifications
  • Compartments:
    -
  • For:
    Tissue culture tubes 15/50 ml
  • W×D×H ext.:
    122×122×128 mm
  • Description:
    without grid insert
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Compartments
    For tubes
    Availability
    Price
  • Grid insert for cryo boxes
    Grid insert for cryo boxes
    NEOL2-7096
    5×5
    15 ml
    Log in for availability
    1 * 1 ST - SFr. 2.20

Product Information

  • Catalog #
    Compartments
    For tubes
    Availability
    Price
  • Grid insert for cryo boxes
    Grid insert for cryo boxes
    NEOL2-7098
    3×3
    50 ml
    Log in for availability
    1 * 1 ST - SFr. 2.60

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.