Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Buffer Solution pH 4.00 (20°C) (red colour) 1 * 1.000 mL
Catalog # APPC273616.1211
Supplier:
PanReac AppliChem
Expand image
Buffer Solution pH 4.00 (20°C) (red colour) 1 * 1.000 mL
Catalog # APPC273616.1211
Supplier:
PanReac AppliChem
Related Information
Documentation
Datasheet
APPC273616.1211_TDS_EN
Documentation
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Ammonium standard solution, 1000 mg/L NH₄ in water (from NH₄Cl) Certipur® standard for Ion Chromatography, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Fluoride standard solution, 1,000 mg/L F- in Water (from NaF) Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Buffer Solution pH 7.00 (20°C) (yellow colour) 1 * 1.000 mL
Supplier: PanReac AppliChem
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach