Buffer Solution pH 4.00 (20°C) (red colour) 1 * 250 mL
Catalog # APPC273616.1209
Supplier: PanReac AppliChem
Buffer Solution pH 4.00 (20°C) (red colour) 1 * 250 mL
Catalog # APPC273616.1209
Supplier: PanReac AppliChem
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Sucrose, Reference Standard
Supplier: USP
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
pH Standard Solutions, IUPAC, Certified
Supplier: Hach