Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Buffers
pH Calibration Buffers
pH buffer solutions
pH buffer solutions
Print
pH buffer solutions
7,00, 20 °C, 250 mL
Catalog # APPC272170.1209
Certificates
Supplier:
PanReac AppliChem
pH buffer solutions
Catalog # APPC272170.1209
Supplier:
PanReac AppliChem
CAS Number:
Certificates
Expand image
Return to Top
pH buffer solutions
7,00, 20 °C, 250 mL
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
SFr. 28.70
Log in
for availability
Unit of Measure
1 * 250 mL
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
Datasheet
APPC272170.1209_TDS_EN
Specifications
Specifications
Pk:
250 mL
Reference temperature:
20 °C
pH value:
7,00
Specifications
Test kits, photometric, AccuVac®
#: 705-0150
Supplier: Hach
VWR® Screw Caps for Bottles, Wide Neck
#: 215-5647
Supplier: VWR Collection