EDTA Disodium Salt 2-hydrate (Reag. Ph. Eur.) for analysis, ACS 1 * 250 g
APPC131669.1209
EDTA Disodium Salt 2-hydrate (Reag. Ph. Eur.) for analysis, ACS 1 * 250 g
APPC131669.1209
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Cover Slips, Round
: VWR Collection
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Charcoal activated, powder (total ash max 4%)
: Thermo Fisher Scientific