Sodium Hydrogen Carbonate for analysis, ACS, ISO 1 * 1.000 g
Catalog # APPC131638.1211
Supplier: PanReac AppliChem
Sodium Hydrogen Carbonate for analysis, ACS, ISO 1 * 1.000 g
Catalog # APPC131638.1211
Supplier: PanReac AppliChem
Frequently Bought Together
Ammonium carbonate, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium carbonate, anhydrous ≥99% ACS
Supplier: VWR Chemicals
Citric acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Aluminium ≥90% stabilised, fine powder, Supelco®
Supplier: Merck
Calcium hydroxide ≥96%, Puriss. p.a. Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Potassium carbonate, anhydrous 98%, extra pure
Supplier: Thermo Fisher Scientific
Tetrabutylammonium hexafluorophosphate, Supelco®
Supplier: Merck
Sodium gluconate 98%
Supplier: Thermo Fisher Scientific
Manganese(II) chloride tetrahydrate 99+% for analysis
Supplier: Thermo Fisher Scientific
di-Potassium oxalate monohydrate ≥99.0% (RT), Purum p.a.
Supplier: Honeywell Chemicals