Sodium Carbonate anhydrous (Reag. Ph. Eur.) for analysis, ACS 1 * 500 g
Catalog # APPC131648.1210
Supplier: PanReac AppliChem
Sodium Carbonate anhydrous (Reag. Ph. Eur.) for analysis, ACS 1 * 500 g
Catalog # APPC131648.1210
Supplier: PanReac AppliChem
Frequently Bought Together
Potassium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Copper(II) sulphate pentahydrate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ammonium iron(III) sulphate dodecahydrate 99.0-101.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium salicylate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals