Sulfuric Acid 1 mol/l (2N) volumetric solution 1 * 1 L
Catalog # APPC182105.1211
Supplier: PanReac AppliChem
Sulfuric Acid 1 mol/l (2N) volumetric solution 1 * 1 L
Catalog # APPC182105.1211
Supplier: PanReac AppliChem
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Disodium hydrogen phosphate dihydrate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
n-Heptane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Urea for biochemistry, Millipore®
Supplier: Merck
Magnesium oxide light 98.0-100.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium iodate, EMSURE® for analysis, Supelco®
Supplier: Merck
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
Thermal transfer ribbon for printer, R6400 series
Supplier: Brady
NANOCOLOR® Photometric Tube Test Kit for Potassium
Supplier: MACHEREY-NAGEL
EDTA disodium salt solution 0.1 mol/l (0.1M) volumetric solution
Supplier: PanReac AppliChem
Carboy res 10L 4-port plus drain 1 * 1 items
Supplier: Spectrum Laboratories
tri-Sodium Citrate 2-hydrate for analysis, ACS 1 * 500 g
Supplier: PanReac AppliChem