Ammonium Chloride for analysis, ACS, ISO 1 * 500 g
Catalog # APPC131121.1210
Supplier: PanReac AppliChem
Ammonium Chloride for analysis, ACS, ISO 1 * 500 g
Catalog # APPC131121.1210
Supplier: PanReac AppliChem
Frequently Bought Together
Magnesium sulphate, EMSURE® for analysis, Supelco®
Supplier: Merck
Magnesium sulphate heptahydrate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Potassium chloride 99.0-100.5% USP, Ph. Eur.
Supplier: VWR Chemicals
di-Potassium hydrogen phosphate ≥99.0% analytical reagent
Supplier: VWR Chemicals
Disodium hydrogen phosphate dodecahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Citric acid, anhydrous ≥99.5%, powder, GPR RECTAPUR®
Supplier: VWR Chemicals
Calcium chloride dihydrate 99.0-103.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium dihydrogen phosphate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium carbonate ≥97%, TECHNICAL
Supplier: VWR Chemicals
Sodium hydrogen carbonate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ammonium acetate ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium acetate, anhydrous 99%
Supplier: Thermo Fisher Scientific
Hydrochloric acid 1 mol/l (1N) volumetric solution
Supplier: PanReac AppliChem