Sulfuric Acid 96% for analysis, ISO 1 * 1.000 mL
Katalognr. APPC131058.1611
Anbieter: PanReac AppliChem
Sulfuric Acid 96% for analysis, ISO 1 * 1.000 mL
Katalognr. APPC131058.1611
Anbieter: PanReac AppliChem
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Anbieter: VWR Chemicals
Bulb pipettes (one mark), BLAUBRAND®, class AS
Anbieter: Brand
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Anbieter: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals