Perchloric Acid 60% for analysis, ACS, ISO 1 * 1.000 mL
Catalog # APPC131054.1611
Supplier: PanReac AppliChem
Perchloric Acid 60% for analysis, ACS, ISO 1 * 1.000 mL
Catalog # APPC131054.1611
Supplier: PanReac AppliChem
Frequently Bought Together
Pyridine ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dimethyl sulphoxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Isobutanol 99+% for HPLC
Supplier: Thermo Fisher Scientific
n-Pentane for analysis
Supplier: Thermo Fisher Scientific
Petroleum spirit, 30…50 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Dodecamolybdophosphoric acid hydrate, Supelco®
Supplier: Merck
n-Hexane ≥95%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
3-(Chloromethyl)benzoyl chloride 98%
Supplier: Thermo Fisher Scientific
FISHER CHEMICAL ACETONITRIL UHPLC GRADIENT GRAD
Supplier: MARKET SOURCE PART PROCESS
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific