SINO HUMAN MFG-E8/ LACTADHERIN MFGE8 PROTEIN (HIS TAG) 1 * 100 µG
Catalog # SINO10853-H08B-100
Supplier: Sino Biological
SINO HUMAN MFG-E8/ LACTADHERIN MFGE8 PROTEIN (HIS TAG) 1 * 100 µG
Catalog # SINO10853-H08B-100
Supplier: Sino Biological
Frequently Bought Together
MicroWell™ Plates, 96 well, Nunc-Immuno™
Supplier: Thermo Fisher Scientific
Transfer pipettes
Supplier: RATIOLAB
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Mouse recombinant TNF alpha (soluble) (from <i>E. coli</i>)
Supplier: ENZO LIFE SCIENCES