Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Viscosity Standards(cSt) reads 790@20C, 580@25C,
[email protected]
, 250@40C, 160@50C 1 * 500 mL
Catalog # REGEREVIS-N250
Supplier:
REAGECON
Expand image
Viscosity Standards(cSt) reads 790@20C, 580@25C,
[email protected]
, 250@40C, 160@50C 1 * 500 mL
Catalog # REGEREVIS-N250
Supplier:
REAGECON
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
pH buffer solutions
Supplier: Mettler - Toledo
S-Pak® Membrane Filters
Supplier: Merck