Multi-element standards, common elements mix 2, VHG
Common elements mix 2, 100 mg/L, 5% HCl/tr HF, 500 ml, 500 mL
Catalog # PROOVHG-SM70B-500
Supplier: LGC Standards PROMOCHEM
Multi-element standards, common elements mix 2, VHG
Catalog # PROOVHG-SM70B-500
Supplier: LGC Standards PROMOCHEM
CAS Number:
Jump to:
Product Details & Documents
This standard solution is intended for use as a certified reference material and calibration standard for inductively coupled plasma mass spectrometry (ICP-MS) or alternative techniques, such as inductively coupled plasma optical emission spectroscopy (ICP-OES), and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Multi elements standards for Ag, Al, B, Ca, Co, Cr, Cu, Fe, K, Mg, Mn, Na, Ni, P, Pb, Si, Sn, Ti, V and Zn.
Certifications: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.
Specifications
Specifications
- Concentration:100 mg/L
- Matrix:5% HCl/tr HF
- Pk:500 mL
- Description:Common elements mix 2
- Pk Info:500 ml



