Orders LinkContactUsLinkComponent
Supelco® Spectroquant® Test Kits, Phosphate
Supelco® Spectroquant® Test Kits, Phosphate
Phosphate cell test*7-3, 0,05-5,0 mg/L PO₄-P; 0,2-15,3 mg/L PO₄; 0,11-11,46 mg/L P₂O₅, Phosphormolybdenum blue, Prove, Nova 30/60, Multy, Move, 25 Tests
Catalog # 1.00474.0001
Supplier:   Merck
Supelco® Spectroquant® Test Kits, Phosphate
Catalog # 1.00474.0001
Supplier:  Merck
CAS Number:  
Supelco® Spectroquant® Test Kits, Phosphate
Supelco® Spectroquant® Test Kits, Phosphate
Phosphate cell test*7-3, 0,05-5,0 mg/L PO₄-P; 0,2-15,3 mg/L PO₄; 0,11-11,46 mg/L P₂O₅, Phosphormolybdenum blue, Prove, Nova 30/60, Multy, Move, 25 Tests
CAS Number:  
Price
SFr. 286.00
Log in for availability
1 * 25 Tests
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*7 This method is officially recognised by the USEPA as an alternative method for the investigation of the following:
*7-1 wastewater
*7-2 drinking water
*7-3 respectively drinking water and waste water
Specifications
Specifications
  • Chemical method:
    Phosphormolybdenum blue
  • Pk:
    25 Tests
  • Description:
    Phosphate cell test*<sup>7-3</sup>
  • For unit:
    Prove, Nova 30/60, Multy, Move
  • Range:
    0,05-5,0 mg/L PO₄-P; 0,2-15,3 mg/L PO₄; 0,11-11,46 mg/L P₂O₅