Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Shelving units

Shelving units

4 solid shelves, on casters, Stainless steel 304, 1000×400×1900 mm
Catalog # 132-0999
Supplier:   Palbam
Shelving units
Catalog # 132-0999
Supplier:  Palbam
Shelving units
Shelving units

4 solid shelves, on casters, Stainless steel 304, 1000×400×1900 mm

Price
4 060.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

High-quality cleanroom shelving units on leveller legs with height adjustable feet or on white nylon 130 mm (5") cleanroom casters.

  • Shelf options: Solid or perforated
  • Shelves are removable or height adjustable every 200 mm (8")
  • 4 shelves per unit (extra shelves can be ordered separately)

Stainless steel 304, electropolished.

Specifications
  • Description:
    4 solid shelves, on casters
  • Material:
    Stainless steel 304
  • Seq:
    3
  • W×D×H:
    1000×400×1900 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.