Hydrochloric Acid 37 % (E-507, F.C.C.) food grade 1 * 25 L
Catalog # APPC201020.0716
Supplier: PanReac AppliChem
Hydrochloric Acid 37 % (E-507, F.C.C.) food grade 1 * 25 L
Catalog # APPC201020.0716
Supplier: PanReac AppliChem
Frequently Bought Together
ASSAY VOLLSTÄNDIGE ZELLE ABLÖSUNG REAGEN
Supplier: DISCOVERX
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
Supplier: PanReac AppliChem
Liquid for manometer with sloping tube
Supplier: VWR Chemicals
tert-Butyl methyl ether for analysis
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Tetrachloroethylene ≥99%
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific