Quarz glass tray in application temperatures of up to 1050°C (maximum 1600°C), 3-sided, with open end, translucent, high chemical resistance and purity. Length 175mm, width 117mm, height 25mm. 1 * 2 items
Catalog # TSLA73042
Supplier: TECHNICAL QUARTZ SOLUTIONS
Frequently Bought Together