Corning® Trypsin
Product Details & Documents
0,05% Trypsin/0,53 mM EDTA in HBSS (Hank’s Balanced Salt Solution) with sodium bicarbonate, without calcium and magnesium.
- Trypsin EDTA 1X 0,05% Trypsin/0,53 mM EDTA in HBSS with sodium bicarbonate, without calcium and magnesium
- Porcine Parvovirus tested
Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging. Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin. This product contains trypsin at a concentration of 0,05% with 0,53 mM EDTA, and does not contain sodium bicarbonate.
Documents
Specifications
Specifications
- Pk:6
- Pk Info:500 ml
- Cat. No.:MDTC25-052-CV
- Enzyme name:Trypsin
- CAS No.:9002-07-7
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



